Merck
  • 2B-Trans technology: targeted drug delivery across the blood-brain barrier.
2B-Trans technology: targeted drug delivery across the blood-brain barrier.
Methods in molecular biology (Clifton, N.J.) (2008-03-29)
Pieter J Gaillard, Albertus G de Boer
ABSTRACT
Drug delivery across the blood-brain barrier (BBB) is a major obstacle for the development of effective treatments of many central nervous system disorders. Sophisticated cell culture models of the BBB have helped us to identify, characterize, and validate a novel targeted drug delivery technology, designated 2B-Trans, for the receptor-mediated uptake and transport of drugs across the BBB. This paper describes in great detail how such a BBB cell culture model should be prepared and handled, and applied for the use of targeted drug delivery across the BBB.
Sigma-Aldrich
Ro-20-1724, A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC₅₀ = 2 µM).