Merck
H3K27me3 conditions chemotolerance in triple-negative breast cancer.
Nature genetics (2022-04-13)
Justine Marsolier, Pacôme Prompsy, Adeline Durand, Anne-Marie Lyne, Camille Landragin, Amandine Trouchet, Sabrina Tenreira Bento, Almut Eisele, Sophie Foulon, Léa Baudre, Kevin Grosselin, Mylène Bohec, Sylvain Baulande, Ahmed Dahmani, Laura Sourd, Eric Letouzé, Anne-Vincent Salomon, Elisabetta Marangoni, Leïla Perié, Céline Vallot
ABSTRACT
The persistence of cancer cells resistant to therapy remains a major clinical challenge. In triple-negative breast cancer, resistance to chemotherapy results in the highest recurrence risk among breast cancer subtypes. The drug-tolerant state seems largely defined by nongenetic features, but the underlying mechanisms are poorly understood. Here, by monitoring epigenomes, transcriptomes and lineages with single-cell resolution, we show that the repressive histone mark H3K27me3 (trimethylation of histone H3 at lysine 27) regulates cell fate at the onset of chemotherapy. We report that a persister expression program is primed with both H3K4me3 (trimethylation of histone H3 at lysine 4) and H3K27me3 in unchallenged cells, with H3K27me3 being the lock to its transcriptional activation. We further demonstrate that depleting H3K27me3 enhances the potential of cancer cells to tolerate chemotherapy. Conversely, preventing H3K27me3 demethylation simultaneously to chemotherapy inhibits the transition to a drug-tolerant state, and delays tumor recurrence in vivo. Our results highlight how chromatin landscapes shape the potential of cancer cells to respond to initial therapy.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
GSK-J4, ≥98% (HPLC)
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Dispase® II, protease
Roche
DNase I, from bovine pancreas