Merck
  • Nickel-catalyzed, sodium iodide-promoted reductive dimerization of alkyl halides, alkyl pseudohalides, and allylic acetates.
Nickel-catalyzed, sodium iodide-promoted reductive dimerization of alkyl halides, alkyl pseudohalides, and allylic acetates.
Chemical communications (Cambridge, England) (2010-06-29)
Michael R Prinsell, Daniel A Everson, Daniel J Weix
ABSTRACT
The first general method for the reductive dimerization of alkyl halides, alkyl mesylates, alkyl trifluoroacetates, and allylic acetates is reported which proceeds with low catalyst loading (0.5 to 5 mol%), generally high yields (80% ave yield), and good functional-group tolerance.
Sigma-Aldrich
4,4′,4″-Tri-tert-Butyl-2,2′:6′,2″-terpyridine, 95%