Merck
  • The epigenetic reader PHF21B modulates murine social memory and synaptic plasticity-related genes.
The epigenetic reader PHF21B modulates murine social memory and synaptic plasticity-related genes.
JCI insight (2022-07-23)
Eunice Wm Chin, Qi Ma, Hongyu Ruan, Camille Chin, Aditya Somasundaram, Chunling Zhang, Chunyu Liu, Martin D Lewis, Melissa White, Tracey L Smith, Malcolm Battersby, Wei-Dong Yao, Xin-Yun Lu, Wadih Arap, Julio Licinio, Ma-Li Wong
ABSTRACT
Synaptic dysfunction is a manifestation of several neurobehavioral and neurological disorders. A major therapeutic challenge lies in uncovering the upstream regulatory factors controlling synaptic processes. Plant homeodomain (PHD) finger proteins are epigenetic readers whose dysfunctions are implicated in neurological disorders. However, the molecular mechanisms linking PHD protein deficits to disease remain unclear. Here, we generated a PHD finger protein 21B-depleted (Phf21b-depleted) mutant CRISPR mouse model (hereafter called Phf21bΔ4/Δ4) to examine Phf21b's roles in the brain. Phf21bΔ4/Δ4 animals exhibited impaired social memory. In addition, reduced expression of synaptic proteins and impaired long-term potentiation were observed in the Phf21bΔ4/Δ4 hippocampi. Transcriptome profiling revealed differential expression of genes involved in synaptic plasticity processes. Furthermore, we characterized a potentially novel interaction of PHF21B with histone H3 trimethylated lysine 36 (H3K36me3), a histone modification associated with transcriptional activation, and the transcriptional factor CREB. These results establish PHF21B as an important upstream regulator of synaptic plasticity-related genes and a candidate therapeutic target for neurobehavioral dysfunction in mice, with potential applications in human neurological and psychiatric disorders.
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GluR1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, Trial Size, Upstate®, from rabbit
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse