Merck
  • Disruption of imprinting and aberrant embryo development in completely inbred embryonic stem cell-derived mice.
Disruption of imprinting and aberrant embryo development in completely inbred embryonic stem cell-derived mice.
Development, growth & differentiation (2007-07-20)
Tao Deng, Ying Kuang, Dan Zhang, Long Wang, Ruilin Sun, Guojiang Xu, Zhugang Wang, Jian Fei
ABSTRACT
The completely embryonic stem (ES) cell-derived mice (ES mice) produced by tetraploid embryo complementation provide us with a rapid and powerful approach for functional genome analysis. However, inbred ES cell lines often fail to generate ES mice. The genome of mouse ES cells is extremely unstable during in vitro culture and passage, and the expression of the imprinted genes is most likely to be affected. Whether the ES mice retain or repair the abnormalities of the donor ES cells has still to be determined. Here we report that the inbred ES mice were efficiently produced with the inbred ES cell line (SCR012). The ES fetuses grew more slowly before day 17.5 after mating, but had an excessive growth from day 17.5 to birth. Five imprinted genes examined (H19, Igf2, Igf2r, Peg1, Peg3) were expressed abnormally in ES fetuses. Most remarkably, the expression of H19 was dramatically repressed in the ES fetuses through the embryo developmental stage, and this repression was associated with abnormal biallelic methylation of the H19 upstream region. The altered methylation pattern of H19 was further demonstrated to have arisen in the donor ES cells and persisted on in vivo differentiation to the fetal stage. These results indicate that the ES fetuses did retain the epigenetic alterations in imprinted genes from the donor ES cells.
Sigma-Aldrich
PluriStem 129/S6 Murine ES cells, The PluriStem 129S6 Murine ES cell line is derived from the widely used 129/S6/SvEv strain of mice (at 3.5 days p.c.) & is intended for injection into black or albino host blastocysts.