Merck
  • Bioconversion of Pinoresinol Diglucoside from Glucose Using Resting and Freeze-Dried Phomopsis sp. XP-8 Cells.
Bioconversion of Pinoresinol Diglucoside from Glucose Using Resting and Freeze-Dried Phomopsis sp. XP-8 Cells.
Journal of microbiology and biotechnology (2017-06-18)
Zhenhong Gao, Muhammad Riaz Rajoka, Jing Zhu, Zhiwei Zhang, Yan Zhang, Jinxin Che, Xiaoguang Xu, Junling Shi
ABSTRACT
Phomopsis sp. XP-8 (an endophytic fungus) was previously found to produce pinoresinol diglucoside (PDG), a major antihypertensive compound of Tu-Chung (the bark of Eucommia ulmoides Oliv.), which is widely used in Chinese traditional medicines. In the present study, two bioconversion systems were developed for the production of PDG in Tris-HCl buffer containing glucose and Phomopsis sp. XP-8 cells (both resting and freeze-dried). When other factors remained unchanged, the bioconversion time, glucose concentration, cell ages, cell dosage, pH, temperature, and stirring speed influenced PDG production in a similar and decreasing manner after an initial increase with increasing levels for each factor. Considering the simultaneous change of various factors, the optimal conditions for PDG production were established as 70 g/l cells (8-day-old), 14 g/l glucose, 28°C, pH 7.5, and 180 rpm for systems employing resting cells, and 3.87 g/l cells, 14.67 g/l glucose, 28°C, pH 7.5, and 180 rpm for systems employing freeze-dried cells. The systems employing freeze-dried cells showed lower peak PDG production (110.28 μg/l), but at a much shorter time (12.65 h) compared with resting cells (23.62 mg/l, 91.5 h). The specific PDG production levels were 1.92 and 24 μg per gram cells per gram glucose for freeze-dried cells and resting cells, respectively. Both systems indicated a new and potentially efficient way to produce PDG independent of microbial cell growth.
Sigma-Aldrich
Pinoresinol diglucoside, ≥98% (HPLC)