Merck
  • Synthesis of a HyCoSuL peptide substrate library to dissect protease substrate specificity.
Synthesis of a HyCoSuL peptide substrate library to dissect protease substrate specificity.
Nature protocols (2017-09-22)
Marcin Poreba, Guy S Salvesen, Marcin Drag
ABSTRACT
Many biologically and chemically based approaches have been developed to design highly active and selective protease substrates and probes. It is, however, difficult to find substrate sequences that are truly selective for any given protease, as different proteases can demonstrate a great deal of overlap in substrate specificities. In some cases, better enzyme selectivity can be achieved using peptide libraries containing unnatural amino acids such as the hybrid combinatorial substrate library (HyCoSuL), which uses both natural and unnatural amino acids. HyCoSuL is a combinatorial library of tetrapeptides containing amino acid mixtures at the P4-P2 positions, a fixed amino acid at the P1 position, and an ACC (7-amino-4-carbamoylmethylcoumarin) fluorescent tag occupying the P1' position. Once the peptide is recognized and cleaved by a protease, the ACC is released and produces a readable fluorescence signal. Here, we describe the synthesis and screening of HyCoSuL for human caspases and legumain. We also discuss possible modifications and adaptations of this approach that make it a useful tool for developing highly active and selective reagents for a wide variety of proteolytic enzymes. The protocol can be divided into three major parts: (i) solid-phase synthesis of the fluorescence-labeled HyCoSuL, (ii) screening of protease P4-P2 preferences, and (iii) synthesis of the optimized activity probes equipped with an AOMK (acyloxymethyl ketone) reactive group and a biotin label for easy detection. Beginning with the library design, the entire protocol can be completed in 4-8 weeks (HyCoSuL synthesis: 3-5 weeks; HyCoSuL screening per enzyme: 4-8 d; and activity-based probe synthesis: 1-2 weeks).
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%