Merck
Emerging therapeutic strategies to enhance HDL function.
Lipids in health and disease (2011-10-12)
Santiago Redondo, José Martínez-González, Concha Urraca, Teresa Tejerina
ABSTRACT
Epidemiologic studies indicate a strong inverse correlation between plasma levels of high-density lipoproteins (HDL) and cardiovascular disease (CVD). The most relevant cardioprotective mechanism mediated by HDL is thought to be reverse cholesterol transport (RCT). New insights in HDL biology and RCT have allowed the development of promising agents aimed to increase HDL function and promote atherosclerosis regression. In this regard, apo-AI analogs and CETP inhibitors dalcetrapib and anacetrapib have aroused a great interest and opened new expectations in the treatment of CVD.
Sigma-Aldrich
Lipoprotein, high density from human plasma, ≥95% (SDS-PAGE), solution
Pricing and availability is not currently available.