Merck
  • Substrate specificity of SIRT1-catalyzed lysine Nepsilon-deacetylation reaction probed with the side chain modified Nepsilon-acetyl-lysine analogs.
Substrate specificity of SIRT1-catalyzed lysine Nepsilon-deacetylation reaction probed with the side chain modified Nepsilon-acetyl-lysine analogs.
Bioorganic chemistry (2009-11-17)
Nuttara Jamonnak, Brett M Hirsch, Yi Pang, Weiping Zheng
ABSTRACT
Peptides containing L-N(epsilon)-acetyl-lysine (L-AcK) or its side chain modified analogs were prepared and assayed using SIRT1, the prototypical human silent information regulator 2 (Sir2) enzyme. While previous studies showed that the side chain acetyl group of L-AcK can be extended to bulkier acyl groups for Sir2 (including SIRT1)-catalyzed lysine N(epsilon)-deacylation reaction, our current study suggested that SIRT1-catalyzed deacetylation reaction had a very stringent requirement for the distance between the alpha-carbon and the side chain acetamido group, with that found in L-AcK being optimal. Moreover, our current study showed that SIRT1 catalyzed the stereospecific deacetylation of L-AcK versus its D-isomer. The results from our current study shall constitute another piece of important information to be considered when designing inhibitors for SIRT1 and Sir2 enzymes in general.
Sigma-Aldrich
Nε-Acetyl-L-lysine