MilliporeSigma
  • Relationship between calcification, atherosclerosis and matrix proteins in the human aorta.
Relationship between calcification, atherosclerosis and matrix proteins in the human aorta.
Folia histochemica et cytobiologica (2021-02-10)
Aleksandra Kuzan, Jerzy Wisniewski, Krzysztof Maksymowicz, Magdalena Kobielarz, Andrzej Gamian, Agnieszka Chwilkowska
ABSTRACT
Extracellular matrix (ECM) proteins have been associated with atherosclerotic complications, such as plaque rupture, calcification and aneurysm. It is not clear what role different types of collagen play in the pathomechanism of atherosclerosis. The aim of the study was to analyze the content of elastin and major types of collagen in the aortic wall and how they associated are with course of atherosclerosis. In this work we present six biochemical parameters related to ECM proteins and collagen-specific amino acids (collagen type I, III, and IV, elastin, proline and hydroxyproline) analyzed in 106 patients' aortic wall specimens characterized by different degree of atherosclerosis. Liquid Chromatography Electrospray Ionization Tandem Mass Spectrometry (LC/ESI-MS/MS), ELISA and immunohistochemical methods were used. The severity of atherosclerosis was assessed on the six-point scale of the American Heart Association, taking into account the number and location of foam cells, the presence of a fatty core, calcium deposits and other characteristic atherosclerotic features. The results show that there is a relationship between the content of collagen-specific amino acids and development of atherosclerosis. The degree of atherosclerotic lesions was negatively correlated with the content of proline, hydroxyproline and the ratio of these two amino acids. Calcium deposits and surrounding tissue were compared and it was demonstrated that the ratio of type I collagen to type III collagen was higher in the aortic tissue than in aortic calcification areas, while the ratio of collagen type III to elastin was smaller in the artery than in the calcium deposits. We suggest that increase in collagen type III presence in the calcification matrix may stem from disorders in the structure of the type I and III collagen fibers. These anomalous fibers are likely to favor accumulation of the calcium salts, an important feature of the process of atheromatosis.
Sigma-Aldrich
Monoclonal Anti-Collagen, Type IV antibody produced in mouse, clone COL-94, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-Collagen, Type III antibody produced in mouse, clone FH-7A, ascites fluid