Merck
  • [Activities of some enzymes and concentration of ammonia in serum of rats with fluoride hyperglycemia].
[Activities of some enzymes and concentration of ammonia in serum of rats with fluoride hyperglycemia].
Annales Academiae Medicae Stetinensis (2006-08-09)
Ewa Grucka-Mamczar, Ewa Birkner, Barbara Stawiarska-Pieta, Hanna Duliban, Renata Polaniak, Dariusz Chlubek, Dorota Samujło, Alicja Cegłowska
ABSTRACT
The study was done in 30 one-month-old Wistar FL rats divided into one control and two study groups of ten animals each. Hyperglycemia was induced with sodium fluoride in water at a concentration of 50 or 100 mg/L during four months. Control animals received distilled water. We observed significantly (p < 0.05) reduced activities of aspartate aminotransferase (by 22.8%) and malic dehydrogenase (by 10.9%) in the group exposed to 100 mg F(-)/L. No pathological changes were revealed in the pancreas of exposed animals.