Merck
  • Reversible cardiac disease features in an inducible CUG repeat RNA-expressing mouse model of myotonic dystrophy.
Reversible cardiac disease features in an inducible CUG repeat RNA-expressing mouse model of myotonic dystrophy.
JCI insight (2021-01-27)
Ashish N Rao, Hannah M Campbell, Xiangnan Guan, Tarah A Word, Xander Ht Wehrens, Zheng Xia, Thomas A Cooper
ABSTRACT
Myotonic dystrophy type 1 (DM1) is caused by a CTG repeat expansion in the DMPK gene. Expression of pathogenic expanded CUG repeat (CUGexp) RNA causes multisystemic disease by perturbing the functions of RNA-binding proteins, resulting in expression of fetal protein isoforms in adult tissues. Cardiac involvement affects 50% of individuals with DM1 and causes 25% of disease-related deaths. We developed a transgenic mouse model for tetracycline-inducible and heart-specific expression of human DMPK mRNA containing 960 CUG repeats. CUGexp RNA is expressed in atria and ventricles and induced mice exhibit electrophysiological and molecular features of DM1 disease, including cardiac conduction delays, supraventricular arrhythmias, nuclear RNA foci with Muscleblind protein colocalization, and alternative splicing defects. Importantly, these phenotypes were rescued upon loss of CUGexp RNA expression. Transcriptome analysis revealed gene expression and alternative splicing changes in ion transport genes that are associated with inherited cardiac conduction diseases, including a subset of genes involved in calcium handling. Consistent with RNA-Seq results, calcium-handling defects were identified in atrial cardiomyocytes isolated from mice expressing CUGexp RNA. These results identify potential tissue-specific mechanisms contributing to cardiac pathogenesis in DM1 and demonstrate the utility of reversible phenotypes in our model to facilitate development of targeted therapeutic approaches.
Sigma-Aldrich
Anti-CUGBP1 Antibody, clone 3B1, clone 3B1, Upstate®, from mouse