Merck
  • Method of transmission filters to measure emission spectra in strongly scattering media.
Method of transmission filters to measure emission spectra in strongly scattering media.
Biomedical optics express (2021-08-31)
Belén Torrado, Alexander Dvornikov, Enrico Gratton
ABSTRACT
We describe a method based on a pair of transmission filters placed in the emission path of a microscope to resolve the emission wavelength of every point in an image. The method can be applied to any type of imaging device that provides the light in the wavelength transmission range of the filters. Unique characteristics of the filter approach are that the light does not need to be collimated and the wavelength response does not depend on the scattering of the sample or tissue. The pair of filters are used to produce the spectral phasor of the transmitted light, which is sufficient to perform spectral deconvolution over a broad wavelength range. The method is sensitive enough to distinguish free and protein-bound NADH and can be used in metabolic studies.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Pricing and availability is not currently available.