Merck
  • Cloning of the ALL-1 fusion partner, the AF-6 gene, involved in acute myeloid leukemias with the t(6;11) chromosome translocation.
Cloning of the ALL-1 fusion partner, the AF-6 gene, involved in acute myeloid leukemias with the t(6;11) chromosome translocation.
Cancer research (1993-12-01)
R Prasad, Y Gu, H Alder, T Nakamura, O Canaani, H Saito, K Huebner, R P Gale, P C Nowell, K Kuriyama
ABSTRACT
Reciprocal chromosome translocations involving 11q23 are frequently associated with acute leukemias, with the t(4;11) translocation predominating among acute lymphoblastic leukemias, and the t(9;11), t(11;19) and t(6;11) translocations most common among acute myeloid leukemias. In each of these translocations the ALL-1 gene, located at 11q23 and constituting the human homologue of Drosophila trithorax, fuses to a specific gene on the partner chromosome to produce a chimeric protein. Here we report the cloning and the characterization of the partner gene from chromosome 6 (AF-6). AF-6 is expressed in a variety of cell types and encodes a protein of 1612 amino acids. The protein contains short stretches rich in prolines, charged amino acids, serines, or glutamines. In addition, the AF-6 protein contains the GLGF motif shared with several proteins of vertebrates and invertebrates thought to be involved in signal transduction at special cell-cell junctions.