Merck
  • Inhibitory effect of highly purified human platelet beta-thromboglobulin on in vitro human megakaryocyte colony formation.
Inhibitory effect of highly purified human platelet beta-thromboglobulin on in vitro human megakaryocyte colony formation.
Experimental hematology (1991-03-01)
J F Abgrall, Z C Han, L Sensebe, G Le Gall, G Guern, I Renard, J Briere
ABSTRACT
The effect of highly purified human beta-thromboglobulin (beta TG), a glycoprotein present in platelet alpha granules, was tested on human bone marrow in vitro megakaryocyte (MK) colony formation. A concentration of 5 micrograms/ml of beta TG induced a 50% reduction in the number of MK colonies, and concentrations of 10 and 20 micrograms/ml completely inhibited MK growth. This inhibition was of importance for MKs because a higher concentration of beta TG (10 micrograms/ml) was needed to obtain a nonsignificant decrease in erythroblastic progenitors (erythroid burst-forming units, BFU-E), and no inhibition of granulocyte-macrophage colony-forming units (CFU-GM) was observed. beta TG acts mainly on maturation of MKs. These results indicate that beta TG could play a role in the physiological regulation of platelet production by MKs.
Sigma-Aldrich
Beta-Thromboglobulin from human platelets, ≥95% (SDS-PAGE)