Merck
  • Regulation of local GTP availability controls RAC1 activity and cell invasion.
Regulation of local GTP availability controls RAC1 activity and cell invasion.
Nature communications (2021-10-21)
Anna Bianchi-Smiraglia, David W Wolff, Daniel J Marston, Zhiyong Deng, Zhannan Han, Sudha Moparthy, Rebecca M Wombacher, Ashley L Mussell, Shichen Shen, Jialin Chen, Dong-Hyun Yun, Anderson O'Brien Cox, Cristina M Furdui, Edward Hurley, Maria Laura Feltri, Jun Qu, Thomas Hollis, Jules Berlin Nde Kengne, Bernard Fongang, Rui J Sousa, Mikhail E Kandel, Eugene S Kandel, Klaus M Hahn, Mikhail A Nikiforov
ABSTRACT
Physiological changes in GTP levels in live cells have never been considered a regulatory step of RAC1 activation because intracellular GTP concentration (determined by chromatography or mass spectrometry) was shown to be substantially higher than the in vitro RAC1 GTP dissociation constant (RAC1-GTP Kd). Here, by combining genetically encoded GTP biosensors and a RAC1 activity biosensor, we demonstrated that GTP levels fluctuating around RAC1-GTP Kd correlated with changes in RAC1 activity in live cells. Furthermore, RAC1 co-localized in protrusions of invading cells with several guanylate metabolism enzymes, including rate-limiting inosine monophosphate dehydrogenase 2 (IMPDH2), which was partially due to direct RAC1-IMPDH2 interaction. Substitution of endogenous IMPDH2 with IMPDH2 mutants incapable of binding RAC1 did not affect total intracellular GTP levels but suppressed RAC1 activity. Targeting IMPDH2 away from the plasma membrane did not alter total intracellular GTP pools but decreased GTP levels in cell protrusions, RAC1 activity, and cell invasion. These data provide a mechanism of regulation of RAC1 activity by local GTP pools in live cells.
Sigma-Aldrich
Anti-IMPDH2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-IMPDH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit