- Some novel antileishmanial compounds inhibit normal cell cycle progression of Leishmania donovani promastigotes and exhibits pro-oxidative potential.
Some novel antileishmanial compounds inhibit normal cell cycle progression of Leishmania donovani promastigotes and exhibits pro-oxidative potential.
In the midst of numerous setbacks that beclouds the fight against leishmaniasis; a neglected tropical disease, the search for new chemotherapeutics against this disease is of utmost importance. Leishmaniasis is a disease closely associated with poverty and endemic in Africa, Asia, southern Europe and the Americas. It is caused by parasites of the genus Leishmania and transmitted by a sandfly vector. In this study, we evaluated the antileishmanial potency of eighteen pathogen box compounds and elucidated their biosafety and possible mechanisms of action against Leishmania donovani promastigotes and amastigotes in vitro. IC50s range of 0.12±0.15 to >6.25 μg/ml and 0.13±0.004 to >6.25μg/ml were observed for the promastigotes and amastigotes, respectively. We demonstrated the ability of some of the compounds to cause cytocidal effect on the parasites, induce increased production of reactive oxygen species (ROS), disrupt the normal parasite morphology and cause the accumulation of parasites at the DNA synthesis phase of the cell cycle. We recommend a further in vivo study on these compounds to validate the findings.