Merck
Insulin-Like Growth Factor Binding Protein-3 Binds to Histone 3.
International journal of molecular sciences (2021-01-07)
Apurva Bhardwaj, Kumar Alok Pathak, Anuraag Shrivastav, Shailly Varma Shrivastav
ABSTRACT
Insulin-like growth factor (IGF) binding protein-3 (IGFBP-3) is an essential protein that regulates cellular processes such as cell proliferation, apoptosis, and differentiation. It is known to bind with several proteins to carry out various cellular functions. In this study, we report for the first time that IGFBP-3 is a histone 3 (H3) binding protein. Sub-cellular fractionation was performed to separate into cytosolic fraction, nucleic acid binding protein fraction and insoluble nuclear fraction. Using ligand blot analysis, we identified a ~15 kDa protein that can interact with IGFBP-3 in the insoluble nuclear fraction. The 15 kDa protein was confirmed as histone 3 by far-Western blot analysis and co-immunoprecipitation experiments. A dot-blot experiment further validated the binding of IGFBP-3 with H3. The intensity of IGFBP-3 on dot-blot showed a proportional increase with H3 concentrations between 2.33 pmol-37.42 pmol. Our results support the presence of protein-protein interaction between IGFBP-3 and H3. The physical binding between IGFBP-3 and H3 could indicate its yet another cellular role in regulating the chromatin remodeling for gene transcription.
Sigma-Aldrich
Histone H3 full length human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
Histone H4 human
Sigma-Aldrich
Histone H2B human
Sigma-Aldrich
Histone H2A human