Merck
  • Oxyresveratrol as the potent inhibitor on dopa oxidase activity of mushroom tyrosinase.
Oxyresveratrol as the potent inhibitor on dopa oxidase activity of mushroom tyrosinase.
Biochemical and biophysical research communications (1998-03-17)
N H Shin, S Y Ryu, E J Choi, S H Kang, I M Chang, K R Min, Y Kim
ABSTRACT
Oxyresveratrol (2,3',4,5'-tetrahydroxystilbene), a naturally occurring compound particularly found in Morus alba L., exhibited a potent inhibitory effect on dopa oxidase activity of tyrosinase which catalyzes rate-limiting steps of melanin biosynthesis. Oxyresveratrol with 0.3 to 5 microM exhibited potent and dose-dependent inhibitions (25 to 84%) on the enzyme activity, where 50% of inhibition was shown at the concentration of about 1 microM. Oxyresveratrol seemed to inhibit the dopa oxidase activity of tyrosinase via a noncompetitive manner (Ki = 9.1 x 10(-7) M) when L-dopa was used as a substrate. Oxyresveratrol exhibited about a 150-fold more potent inhibitory effect than resveratrol (3,4',5-trihydroxystilbene). The more hydroxy groups of the hydroxystilbenes are methylated to be methoxy groups, while the less inhibitory effects on the enzyme activity were exhibited. The results indicate that both the number and positions of hydroxy groups in oxyresveratrol seem to play a critical role in exerting the inhibitory effect on dopa oxidase activity of mushroom tyrosinase.
Sigma-Aldrich
Oxyresveratrol, ≥97.0% (HPLC)