- Combinatorial Treatment of Tinzaparin and Chemotherapy Can Induce a Significant Antitumor Effect in Pancreatic Cancer.
Combinatorial Treatment of Tinzaparin and Chemotherapy Can Induce a Significant Antitumor Effect in Pancreatic Cancer.
Pancreatic Cancer (PC) is recognized as a highly thrombogenic tumor; thus, low-molecular-weight heparin (LMWH) such as tinzaparin is routinely used for PC patients. On the basis of combinatorial therapy approaches to treat highly malignant and refractory cancers such as PC, we hypothesized that tinzaparin can augment the effectiveness of traditional chemotherapeutic drugs and induce efficient antitumor activity. PANC-1 and MIAPaCa-2 were incubated alone or in combination with tinzaparin, nab-paclitaxel and gemcitabine. In vivo evaluation of these compounds was performed in a NOD/SCID mouse using a model injected with PANC-1. Tinzaparin enhances the anti-tumor effects of nab-paclitaxel and gemcitabine in mtKRAS PC cell lines via apoptosis in in vitro experiments. The triple combination power acts through the induction of apoptosis, reduction of the proliferative potential and angiogenesis; hence, contributing to a decrease in tumor volume observed in vivo. The triple regimen provided an extra 24.3% tumor reduction compared to the double combination (gemcitabine plus nab-paclitaxel). Combinatorial strategies can create novel therapeutic approaches for the treatment of patients with PC, achieving a better clinical outcome and prolonged survival. Further prospective randomized research is needed and the investigation of various concentrations of tinzaparin above 150 UI/Kg, would potentially provide a valuable synergistic effect to the conventional therapeutic compounds.