Merck
  • Prenylated Trans-Cinnamic Esters and Ethers against Clinical Fusarium spp.: Repositioning of Natural Compounds in Antimicrobial Discovery.
Prenylated Trans-Cinnamic Esters and Ethers against Clinical Fusarium spp.: Repositioning of Natural Compounds in Antimicrobial Discovery.
Molecules (Basel, Switzerland) (2021-01-31)
Safa Oufensou, Stefano Casalini, Virgilio Balmas, Paola Carta, Wiem Chtioui, Maria A Dettori, Davide Fabbri, Quirico Migheli, Giovanna Delogu
ABSTRACT
Onychomycosis is a common nail infection mainly caused by species belonging to the F. oxysporum, F. solani, and F. fujikuroi species complexes. The aim of this study was to evaluate the in vitro susceptibility of six representative strains of clinically relevant Fusarium spp. toward a set of natural-occurring hydroxycinnamic acids and their derivatives with the purpose to develop naturally occurring products in order to cope with emerging resistance phenomena. By introducing a prenylated chain at one of the hydroxy groups of trans-cinnamic acids 1-3, ten prenylated derivatives (coded 4-13) were preliminarily investigated in solid Fusarium minimal medium (FMM). Minimal inhibitory concentration (MIC) and lethal dose 50 (LD50) values were then determined in liquid FMM for the most active selected antifungal p-coumaric acid 3,3'-dimethyl allyl ester 13, in comparison with the conventional fungicides terbinafine (TRB) and amphotericin B (AmB), through the quantification of the fungal growth. Significant growth inhibition was observed for prenylated derivatives 4-13, evidencing ester 13 as the most active. This compound presented MIC and LD50 values (62-250 µM and 7.8-125 µM, respectively) comparable to those determined for TRB and AmB in the majority of the tested pathogenic strains. The position and size of the prenylated chain and the presence of a free phenol OH group appear crucial for the antifungal activity. This work represents the first report on the activity of prenylated cinnamic esters and ethers against clinical Fusarium spp. and opens new avenues in the development of alternative antifungal compounds based on a drug repositioning strategy.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture