Merck
  • Synthesis of new 2-substituted pyrido[2,3-d]pyrimidin-4(1H)-ones and their antibacterial activity.
Synthesis of new 2-substituted pyrido[2,3-d]pyrimidin-4(1H)-ones and their antibacterial activity.
European journal of medicinal chemistry (2008-07-08)
B Lakshmi Narayana, A Raghu Ram Rao, P Shanthan Rao
ABSTRACT
2-Substituted-5,7-dimethyl pyrido[2,3-d]pyrimidin-4(1H)-ones (8) were synthesized by oxidation of 2-substituted-5,7-dimethyl dihydropyrido[2,3-d]pyrimidin-4(1H)-ones (7) which were in turn prepared from 2-amino-4,6-dimethyl nicotinamide (5) and substituted aryl aldehydes (6). 2-Amino-4,6-dimethyl nicotinamide (5) was prepared from ethyl cyanoacetate (1) via malonamamidine hydrochloride (3). The compounds were characterized by IR, NMR, MS and elemental analyses. Compounds 7 and 8 were screened for antibacterial activity against gram positive and gram negative bacteria. Dehydrogenated compounds (8) showed less antibacterial activity than the compounds 7. Among all the test compounds screened for antibacterial activity 7c (1.25 microg/ml) showed greater activity. All the synthesized compounds were found inactive when screened for antifungal activity at the concentration of 200 microg/ml.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Pricing and availability is not currently available.