Merck
  • Spatio-temporal dynamics of β-tubulin isotypes during the development of the sensory auditory organ in rat.
Spatio-temporal dynamics of β-tubulin isotypes during the development of the sensory auditory organ in rat.
Histochemistry and cell biology (2015-07-27)
Justine Renauld, Nicolas Johnen, Nicolas Thelen, Marie Cloes, Marc Thiry
ABSTRACT
There are different β-tubulin isoforms in microtubules of vertebrate tissues. However, their functional significance is still largely unknown. In the present study, we investigated the localization of five β-tubulin isotypes (β1-5) within the hearing organ during development in rat. By using confocal microscopy, we showed that with the exception of the β3-tubulin isoform that was specific to nerve fibres, all the different β-tubulin isoforms were mainly present in the supporting cells. Contrary to β1-4-tubulins, we also found that the β5-tubulin isoform appeared only at a key stage of the post-natal development in specific cell types (pillar cells and Deiters' cells). By using transmission electron microscopy, we revealed further that this developmental stage coincided with the formation of two separate bundles of microtubules from a unique one in these supporting cells. Together, these results suggest that the β5-tubulin isoform might be involved in the generation of new microtubule bundles from a pre-existing one.
Sigma-Aldrich
Anti-β-Tubulin II antibody, Mouse monoclonal, clone 7B9, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I antibody produced in mouse, clone SAP.4G5, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin Isotype III antibody, Mouse monoclonal, clone SDL.3D10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Myosin VI (KA-15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin IV antibody produced in mouse, clone ONS.1A6, ascites fluid