Merck
Role of meprin A in renal tubular epithelial cell injury.
Kidney international (2007-03-23)
C Herzog, R Seth, S V Shah, G P Kaushal
ABSTRACT
Meprins are zinc-dependent metalloproteinases that are highly expressed in the brush-border membranes of both the kidney and the intestines. Meprins are capable of proteolytically degrading extracellular matrix proteins, proteolytically processing bioactive proteins, and play a role in inflammatory processes. In this study, the function of meprin A in the acute kidney injury (AKI) model of cisplatin nephrotoxicity was examined. Normal linear localization of meprin A in the brush border membranes of proximal tubules was altered in AKI. The meprin A alpha-subunit was detected in the urine of both control and cisplatin-treated mice. A cleaved product of the meprin A beta-subunit, undetected in the urine of control mice, was found to be significantly increased in the urine during the progression of cisplatin nephrotoxicity. The excretion of this beta-fragment was found to be before the rise in serum creatinine and blood urea nitrogen (BUN) suggesting usefulness as a biomarker for AKI. Pretreatment of mice with a meprin A inhibitor afforded protection from cisplatin nephrotoxicity as reflected by significant decreases in serum creatinine, BUN, and the excretion of kidney injury molecule-1. These decreases in serum and urine biomarkers were accompanied by significant decreases in histologic markers such as leukocyte infiltration and apoptosis. Meprin A appears to be an important therapeutic target and urinary excretion appears to be a potential biomarker of AKI.
Sigma-Aldrich
Leukocyte Naphthol AS-D Chloroacetate (Specific Esterase) Kit