Merck
  • The effect of excess weight on circulating inflammatory cytokines in drug-naïve first-episode psychosis individuals.
The effect of excess weight on circulating inflammatory cytokines in drug-naïve first-episode psychosis individuals.
Journal of neuroinflammation (2018-03-02)
María Juncal-Ruiz, Laura Riesco-Dávila, Víctor Ortiz-García de la Foz, Mariluz Ramírez-Bonilla, Obdulia Martínez-García, Juan Irure-Ventura, Juan Carlos Leza, Marcos López-Hoyos, Benedicto Crespo-Facorro
ABSTRACT
Low-grade inflammation has been repeatedly associated with both excess weight and psychosis. However, no previous studies have addressed the direct effect of body mass index (BMI) on basal serum cytokines in individuals with first-episode psychosis (FEP). The aim of this study is to analyze the effect of BMI on basal serum cytokine levels in FEP patients and control subjects, separating the total sample into two groups: normal-weight and overweight individuals. This is a prospective and open-label study. We selected 75 FEP patients and 75 healthy controls with similar characteristics to patients according to the following variables: sex, age, and cannabis and tobacco consumption. Both controls and patients were separated into two groups according to their BMI: subjects with a BMI under 25 were considered as normal weight and those with a BMI equal to or more than 25 were considered as overweight. Serum levels of 21 cytokines/chemokines were measured at baseline using the Human High Sensitivity T Cell Magnetic Bead Panel protocol from the Milliplex® Map Kit. We compared the basal serum levels of the 21 cytokines between control and patient groups according to their BMI. In the normal-weight group, IL-8 was the only cytokine that was higher in patients than in the control group (p = 0.001), whereas in the overweight group, serum levels of two pro-inflammatory cytokines (IL-6, p = 0.000; IL-1β, p = 0.003), two chemokines (IL-8, p = 0.001; MIP-1β, p = 0.001), four Th-1 and Th-2 cytokines (IL-13, p = 0.009; IL-2, p = 0.001; IL-7, p = 0.001; IL-12p70, p = 0.010), and one Type-3 cytokine (IL-23, p = 0.010) were higher in patients than in controls. Most differences in the basal serum cytokine levels between patients and healthy volunteers were found in the overweight group. These findings suggest that excess weight can alter the homeostasis of the immune system and therefore may have an additive pro-inflammatory effect on the one produced by psychosis in the central nervous system.
Product Description
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel Premixed 21-plex - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.