- Spontaneous colitis in IL-10-deficient mice was ameliorated via inhibiting glutaminase1.
Spontaneous colitis in IL-10-deficient mice was ameliorated via inhibiting glutaminase1.
Immunity imbalance and barrier damage in the intestinal mucosa are the main pathogenic factors of Crohn's disease (CD). Bis-2-(5-phenylacetamido-1,2,4-thiadiazol-2-yl) ethyl sulfide (BPTES) is a glutaminase 1 (Gls1) inhibitor with the dual functions of increasing glutamine levels and immune regulation. In this study, we focused on the role of BPTES in CD-like enteritis and the possible mechanisms. We found that Gls1 expression was significantly increased in CD intestinal tissue compared with control tissue. Bis-2-(5-phenylacetamido-1,2,4-thiadiazol-2-yl) ethyl sulfide treatment significantly ameliorated chronic colitis in the IL-10-/- , as manifested by decreased disease activity index, body weight change, histological inflammatory degree and inflammatory cytokine expression. Bis-2-(5-phenylacetamido-1,2,4-thiadiazol-2-yl) ethyl sulfide treatment exerted protective effects on CD that were associated with the maintenance of intestinal barrier integrity and the Th/Treg balance. Bis-2-(5-phenylacetamido-1,2,4-thiadiazol-2-yl) ethyl sulfide treatment may act in part through TCR-mediated mammalian target of rapamycin complex 1 (mTORC1) signalling activation. In conclusion, inhibition of Gls1 expression attenuated chronic colitis by maintaining intestinal barrier integrity and the Th/Treg balance, thereby ameliorating CD-like colitis.