Merck
  • Hybrid selection of discrete genomic intervals on custom-designed microarrays for massively parallel sequencing.
Hybrid selection of discrete genomic intervals on custom-designed microarrays for massively parallel sequencing.
Nature protocols (2009-05-30)
Emily Hodges, Michelle Rooks, Zhenyu Xuan, Arindam Bhattacharjee, D Benjamin Gordon, Leonardo Brizuela, W Richard McCombie, Gregory J Hannon
ABSTRACT
Complementary techniques that deepen information content and minimize reagent costs are required to realize the full potential of massively parallel sequencing. Here, we describe a resequencing approach that directs focus to genomic regions of high interest by combining hybridization-based purification of multi-megabase regions with sequencing on the Illumina Genome Analyzer (GA). The capture matrix is created by a microarray on which probes can be programmed as desired to target any non-repeat portion of the genome, while the method requires only a basic familiarity with microarray hybridization. We present a detailed protocol suitable for 1-2 microg of input genomic DNA and highlight key design tips in which high specificity (>65% of reads stem from enriched exons) and high sensitivity (98% targeted base pair coverage) can be achieved. We have successfully applied this to the enrichment of coding regions, in both human and mouse, ranging from 0.5 to 4 Mb in length. From genomic DNA library production to base-called sequences, this procedure takes approximately 9-10 d inclusive of array captures and one Illumina flow cell run.
Product Description
Roche
Agarose LE, low electroendosmosis
Roche
Rapid DNA Ligation Kit
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)