Merck
Fluorescent indicators for cytosolic sodium.
The Journal of biological chemistry (1989-11-15)
A Minta, R Y Tsien
ABSTRACT
Fluorescent indicators sensitive to cytosolic concentrations of free Na+ have been synthesized and characterized. They consist of a crown ether, 1,7-diaza-4,10,13-trioxacyclopentadecane, linked via its nitrogens to fluorophores bearing additional liganding centers. In the currently preferred dye, SBFI (short for sodium-binding benzofuran isophthalate), the fluorophores are benzofurans linked to isophthalate groups. Selectivities for Na+ over K+ of about 20 are observed, resulting in effective dissociation constants for Na+ of about 20 mM against a background of 120 mM K+. Increasing [Na+] increases the ratio of excitation efficiency at 330-345 nm to that at 370-390 nm with emission collected at 450-550 nm, so that ratio fluorometry and imaging can be performed with the same wavelengths as used with the well known Ca2+ indicator fura-2. If the macrocyclic ring is increased in size to a 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, the chelators become selective for K+ over Na+.
Sigma-Aldrich
PBFI-AM, suitable for fluorescence, ≥98.0% (HPCE)