Merck
  • Are changes in MAPK/ERK necessary or sufficient for entrainment in chick pineal cells?
Are changes in MAPK/ERK necessary or sufficient for entrainment in chick pineal cells?
The Journal of neuroscience : the official journal of the Society for Neuroscience (2003-11-07)
Geetha Yadav, Martin Straume, James Heath, Martin Zatz
ABSTRACT
Chick pineal cells in culture display a circadian rhythm of melatonin release. Light pulses can entrain (phase shift) the rhythm. One candidate for the photoentrainment pathway uses a mitogen-activated protein kinase (MAPK), also known as extracellular signal-regulated kinase (ERK). We tested the hypothesis that activation of ERK (by phosphorylation to p-ERK) is necessary and/or sufficient for entrainment by measuring the ability of several drugs, light, and other perturbations to change levels of p-ERK and to induce phase shifts in the melatonin rhythm. If changes in the levels of p-ERK are sufficient for photoentrainment, then all perturbations that reduce its level must induce light-like phase shifts, and all those that increase its level must induce dark-like phase shifts. If such changes are necessary for photoentrainment, then light pulses must reduce p-ERK levels, and the duration of the light pulse, the magnitude and duration of the change in p-ERK, and the size of the phase shift must correlate. We found five perturbations that reduced p-ERK levels. Of these, two induced light-like phase shifts (PD 98059 and caffeine), one induced dark-like phase shifts (SB203580), and two did not induce phase shifts at all (U0126 and omitting a medium change). Serum increased p-ERK levels without inducing any phase shifts. Finally, light pulses did not elicit changes in p-ERK, nor was there a diurnal rhythm in p-ERK levels, nor could rapid changes in p-ERK levels have accounted for duration effects of light pulses on phase shifts. Taken together, these results argue strongly against the hypothesis that reduction (or increases) in MAPK/ERK activation is necessary or sufficient for entrainment in chick pineal cells.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
SB 203580, Hydrochloride, SB 203580, Hydrochloride, CAS 869185-85-3, is a water-soluble form of SB 203580. A highly potent, selective, ATP-competitive inhibitor of p38 MAP Kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.