Merck
  • Single-Molecule Imaging Reveals Translation of mRNAs Localized to Stress Granules.
Single-Molecule Imaging Reveals Translation of mRNAs Localized to Stress Granules.
Cell (2020-12-15)
Daniel Mateju, Bastian Eichenberger, Franka Voigt, Jan Eglinger, Gregory Roth, Jeffrey A Chao
ABSTRACT
Cellular stress leads to reprogramming of mRNA translation and formation of stress granules (SGs), membraneless organelles consisting of mRNA and RNA-binding proteins. Although the function of SGs remains largely unknown, it is widely assumed they contain exclusively non-translating mRNA. Here, we re-examine this hypothesis using single-molecule imaging of mRNA translation in living cells. Although we observe non-translating mRNAs are preferentially recruited to SGs, we find unequivocal evidence that mRNAs localized to SGs can undergo translation. Our data indicate that SG-associated translation is not rare, and the entire translation cycle (initiation, elongation, and termination) can occur on SG-localized transcripts. Furthermore, translating mRNAs can be observed transitioning between the cytosol and SGs without changing their translational status. Together, these results demonstrate that mRNA localization to SGs is compatible with translation and argue against a direct role for SGs in inhibition of protein synthesis.
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
Universal Food pathogen ID Membrane, light pink colored, sterile, diameter 70 mm, suitable for microbiology
Sigma-Aldrich
Ganciclovir, ≥99% (HPLC), powder