Merck
  • Design of a potent reactivator of tabun-inhibited acetylcholinesterase--synthesis and evaluation of (E)-1-(4-carbamoylpyridinium)-4-(4-hydroxyiminomethylpyridinium)-but-2-ene dibromide (K203).
Design of a potent reactivator of tabun-inhibited acetylcholinesterase--synthesis and evaluation of (E)-1-(4-carbamoylpyridinium)-4-(4-hydroxyiminomethylpyridinium)-but-2-ene dibromide (K203).
Journal of medicinal chemistry (2007-10-11)
Kamil Musilek, Daniel Jun, Jiri Cabal, Jiri Kassa, Frank Gunn-Moore, Kamil Kuca
ABSTRACT
Acetylcholinesterase reactivators are crucial antidotes for the treatment of organophosphate intoxication. Among the organophosphates, with the exception of soman, tabun (GA) intoxications are the least responsive to treatment with commercially available therapeutics. A rational design was used to increase reactivation ability and decrease the toxicity of the novel reactivator. (E)-1-(4-carbamoylpyridinium)-4-(4-hydroxyiminomethylpyridinium)-but-2-ene dibromide (K203) has better properties than previously tested compounds in vitro and, therefore, is a potential candidate for the treatment of GA intoxication in vivo.
Sigma-Aldrich
Pyridine-2-aldoxime methochloride