Merck
  • Characterization of antifungal compounds produced by lactobacilli in cheese-mimicking matrix: Comparison between active and inactive strains.
Characterization of antifungal compounds produced by lactobacilli in cheese-mimicking matrix: Comparison between active and inactive strains.
International journal of food microbiology (2020-08-11)
Meg Rouxel, Manon Barthe, Pierre Marchand, Camille Juin, Leslie Mondamert, Thierry Berges, Philippe Blanc, Julien Verdon, Jean-Marc Berjeaud, Willy Aucher
ABSTRACT
Biopreservation of dairy products by acid lactic bacteria appears as a promising alternative to either replace or reduce the use of chemical preservatives. This study aimed at the identification of bacteria preventing fungal spoilers growth in dairy products, and, at the understanding of their antifungal activity. First, antifungal activity of eighteen Lactobacillus strains was tested against five molds and four yeasts leading to selection of L. casei 7006 which had an activity against seven fungal targets. Then, challenge tests against C. lusistaniae 3668 in a cheese-mimicking matrix have been performed demonstrating that this strain was able to reduce strongly this yeast growth after 14 and 21 days storages at 7 °C. Antifungal compounds produced in cheese-mimicking matrix containing L. casei 7006 strain were quantified, then compared to the one prepared with an inactive strain (L. casei 6960) or without Lactobacillus strain. Three compounds were differently produced between cheeses with or without Lactobacillus strain after 21 days at 7 °C: lactic acid, benzoic acid and diacetyl. However, lactic acid concentrations were similar between the three cheeses after 14 days at 7 °C, but an antifungal activity was only associated to L. casei 7006 presence. Benzoic acid concentrations between cheese with L. casei 7006 and negative control L. casei 6960 were also the same. Among the antifungal molecules retrieved from these analyses, diacetyl was the most significantly overproduced in cheese containing L. casei 7006, thus this volatile was associated to the antifungal activity of this strain.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Supelco
C4 - C24 Even Carbon Saturated FAMEs, 1000 μg/mL each component in hexane, analytical standard
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
F.A.M.E. Mix, C4-C24, certified reference material,  wt. % (varied)
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%