Merck
  • Efficiency comparison of apigenin-7-O-glucoside and trolox in antioxidative stress and anti-inflammatory properties.
Efficiency comparison of apigenin-7-O-glucoside and trolox in antioxidative stress and anti-inflammatory properties.
The Journal of pharmacy and pharmacology (2020-08-04)
Wei Wang, Ru-Feng Yue, Zhen Jin, Li-Min He, Rong Shen, Dan Du, You-Zhi Tang
ABSTRACT
Chamomile has long been used as a medicinal plant due to its antioxidative and anti-inflammatory activity. Apigenin-7-O-glucoside (AG) is one of the major ethanol extract components from chamomile; however, the underlying mechanism remains unclear. In this study, the antioxidant potential and the anti-inflammatory activities of AG were analysed and compared with those of trolox. We demonstrate the protective effects of AG on free radical-induced oxidative damage of DNA, proteins and erythrocytes. Flow cytometry assay was used to detect ROS production. Additionally, the expression of anti-oxidation-related and inflammation-related factors was detected by ELISA and Western blotting, respectively. AG and trolox showed different efficiency as antioxidant in different experimental systems. AG had similar effect as trolox to inhibit H2 O2 -induced ROS production in RAW264.7 cells, while exerted stronger inhibition against free radical-induced oxidative damage on erythrocytes than trolox. Interestingly, compared with trolox, AG also had stronger inhibitory effect on LPS-induced NF-κB/NLRP3/caspase-1 signalling in RAW246.7 cells. These results suggest the potential of AG as a pharmaceutical drug for anti-oxidation and anti-inflammation, and the combined usage of AG and trolox might promote its efficacy. Our findings will provide new insights into the development of new drugs with antioxidative and anti-inflammatory functions.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)