Merck
  • Antifungal chromans inhibiting the mitochondrial respiratory chain of pea seeds and new xanthones from Calophyllum caledonicum.
Antifungal chromans inhibiting the mitochondrial respiratory chain of pea seeds and new xanthones from Calophyllum caledonicum.
Planta medica (2004-01-30)
A-E Hay, D Guilet, C Morel, G Larcher, D Macherel, A-M Le Ray, M Litaudon, P Richomme
ABSTRACT
Two new xanthones, caledonixanthone M 1 and caloxanthone L 2, and one new acid, caledonic acid 6 were isolated from the hexane-soluble extract of the stem bark of Calophyllum caledonicum. In the course of this phytochemical study, seven other known compounds - calothwaitesixanthone, calozeyloxanthone, allanxanthone, isoapetalic acid 3, calolongic acid 4, apetalic acid 5 and isocalolongic acid 7 - were isolated. Their antifungal activity against the growth of the human pathogenic fungus Aspergillus fumigatus was then investigated. The results indicated that the crude extract, calolongic acid 4 and isocalolongic acid 7 exhibited strong inhibitory effects with MIC (80) values of 8, 4, 2 microg/mL, respectively. Besides, calolongic acid 4, its lactone derivative 4a and isocalolongic acid 7 markedly reduced the respiration of pea seed mitochondria.
Sigma-Aldrich
Isocalolongic acid, ≥90% (LC/MS-ELSD)