- Use of Pyrazole Hydrogen Bonding in Tripodal Complexes to Form Self Assembled Homochiral Dimers.
Use of Pyrazole Hydrogen Bonding in Tripodal Complexes to Form Self Assembled Homochiral Dimers.
The 3:1 condensation of 5-methyl-1H-pyrazole-3-carboxaldehyde (MepyrzH) with tris(2-aminoethyl)amine (tren) gives the tripodal ligand tren(MePyrzH)3. Aerial oxidation of a solution of cobalt(II) with this ligand in the presence of base results in the isolation of the insoluble Co(tren)(MePyrz)3. This complex reacts with acids, HCl/NaClO4, NH4ClO4, NH4BF4, and NH4I to give the crystalline compounds Co(tren)(MePyrzH)3(ClO4)3, {[Co(tren)(MePyrzH0.5)3](ClO4)1.5}2 {[Co(tren)(MePyrzH0.5)3](BF4)1.5}2 and [Co(tren)(MePyrzH)3][Co(tren)(MePyrzH)3]I2. The latter three complexes are dimeric, held together by three Npyrazole -H…Npyrazolate hydrogen bonds. The structures and symmetries of these homochiral dimers or pseudodimers are discussed in terms of their space group. Possible applications of these complexes by incorporation into new materials are mentioned.