Merck
  • Interference with SMO increases chemotherapy drug sensitivity of A2780/DDP cells by inhibiting the Hh/Gli signaling pathway.
Interference with SMO increases chemotherapy drug sensitivity of A2780/DDP cells by inhibiting the Hh/Gli signaling pathway.
Journal of cellular biochemistry (2020-01-07)
Shihong Ma, Dan Liu, Wenhua Tan, Botao Du, Wei Liu, Weijia Li, Yufei Jiao
ABSTRACT
Aberrant activation of the Hedgehog (Hh)/Gli pathway contributes to the tumorigenesis of several human cancers, including ovarian cancers. We investigated the function of SMO on cell growth, drug resistance, and invasive ability in A2780/DDP cells. Moreover, we also tested the levels of the downstream target genes of the Hh/Gli pathway in SMO short hairpin RNA (shRNA) lentivirus-infected A2780/DDP cells. Western blot analysis results revealed that the Hh/Gli pathway was activated in cisplatin-resistant A2780/DDP cells. After infection by SMO shRNA lentivirus, the colony formation rate and invasive rate of cisplatin-resistant A2780/DDP cells were decreased. 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay showed that upon transfection with SMO shRNA, cell growth was decreased and drug sensitivity to cisplatin was upregulated. Moreover, interference with SMO decreased the expression of MMP-2, MMP-9, VEGF, and Snail in cisplatin-resistant cells. Thus, the Hh/Gli signaling pathway was aberrantly activated in A2780/DDP cells. The colony formation rate and invasive rate were decreased in SMO shRNA lentivirus-infected A2780/DDP cells. All results showed that inhibiting Hh/Gli signaling may negatively regulate the proliferation, invasion, and metastasis of cisplatin-resistant A2780/DDP cells, as well as increase the sensitivity of A2780/DDP to the chemotherapeutic drug of cisplatin.
Product Description
Cisplatin, British Pharmacopoeia (BP) Reference Standard