Merck
  • NEK7 mediated assembly and activation of NLRP3 inflammasome downstream of potassium efflux in ventilator-induced lung injury.
NEK7 mediated assembly and activation of NLRP3 inflammasome downstream of potassium efflux in ventilator-induced lung injury.
Biochemical pharmacology (2020-05-01)
Huan Liu, Changping Gu, Mengjie Liu, Ge Liu, Yuelan Wang
ABSTRACT
Disordered immune regulation and persistent inflammatory damage are the key mechanisms of ventilator-induced lung injury (VILI). NLR family pyrin domain containing 3 (NLRP3) inflammasome activation causes VILI by mediating the formation of inflammatory mediators and infiltration of inflammatory cells, increasing pulmonary capillary membrane permeability, which leads to pulmonary edema and lung tissue damage. What mediates activation of NLRP3 inflammasome in VILI? In this study, we constructed an in vitro cyclic stretch (CS)-stimulated mouse lung epithelial (MLE-12) cell model that was transfected with NIMA-related kinase 7 (NEK7) small interfering RNA (siRNA) or scramble siRNA (sc siRNA) and pretreated with or without glibenclamide (glb). We also established a VILI mouse model, which was pretreated with glibenclamide or oridonin (Ori). Our goal was to investigate the regulatory effects of NEK7 on NLRP3 inflammasome activation and the anti-inflammatory effects of glibenclamide and oridonin on VILI. Mechanical stretch exaggerated the interaction between NEK7 and NLRP3, leading to assembly and activation of NLRP3 inflammasome downstream of potassium efflux. NEK7 depletion and treatment with glibenclamide or oridonin exerted anti-inflammatory effects that alleviated VILI by blocking the interaction between NEK7 and NLRP3, inhibiting NLRP3 inflammasome activation. NEK7 is a vital mediator of NLRP3 inflammasome activation, and glibenclamide or oridonin may be candidates for the development of new therapeutics against VILI driven by the interaction between NEK7 and NLRP3.
Sigma-Aldrich
MISSION® esiRNA, targeting human NEK7