Merck
First evidence of production of the lantibiotic nisin P.
Scientific reports (2020-03-01)
Enriqueta Garcia-Gutierrez, Paula M O'Connor, Gerhard Saalbach, Calum J Walsh, James W Hegarty, Caitriona M Guinane, Melinda J Mayer, Arjan Narbad, Paul D Cotter
ABSTRACT
Nisin P is a natural nisin variant, the genetic determinants for which were previously identified in the genomes of two Streptococcus species, albeit with no confirmed evidence of production. Here we describe Streptococcus agalactiae DPC7040, a human faecal isolate, which exhibits antimicrobial activity against a panel of gut and food isolates by virtue of producing nisin P. Nisin P was purified, and its predicted structure was confirmed by nanoLC-MS/MS, with both the fully modified peptide and a variant without rings B and E being identified. Additionally, we compared its spectrum of inhibition and minimum inhibitory concentration (MIC) with that of nisin A and its antimicrobial effect in a faecal fermentation in comparison with nisin A and H. We found that its antimicrobial activity was less potent than nisin A and H, and we propose a link between this reduced activity and the peptide structure.
Sigma-Aldrich
Ethidium bromide monoazide, ≥95% (HPLC), solid
Sigma-Aldrich
1,1-Dimethylcyclohexane, 99%