Merck
  • WDR90 is a centriolar microtubule wall protein important for centriole architecture integrity.
WDR90 is a centriolar microtubule wall protein important for centriole architecture integrity.
eLife (2020-09-19)
Emmanuelle Steib, Marine H Laporte, Davide Gambarotto, Natacha Olieric, Celine Zheng, Susanne Borgers, Vincent Olieric, Maeva Le Guennec, France Koll, Anne-Marie Tassin, Michel O Steinmetz, Paul Guichard, Virginie Hamel
ABSTRACT
Centrioles are characterized by a nine-fold arrangement of microtubule triplets held together by an inner protein scaffold. These structurally robust organelles experience strenuous cellular processes such as cell division or ciliary beating while performing their function. However, the molecular mechanisms underlying the stability of microtubule triplets, as well as centriole architectural integrity remain poorly understood. Here, using ultrastructure expansion microscopy for nanoscale protein mapping, we reveal that POC16 and its human homolog WDR90 are components of the microtubule wall along the central core region of the centriole. We further found that WDR90 is an evolutionary microtubule associated protein. Finally, we demonstrate that WDR90 depletion impairs the localization of inner scaffold components, leading to centriole structural abnormalities in human cells. Altogether, this work highlights that WDR90 is an evolutionary conserved molecular player participating in centriole architecture integrity.
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Anti-FGFR1OP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered