Merck
  • Discovery of BI 99179, a potent and selective inhibitor of type I fatty acid synthase with central exposure.
Discovery of BI 99179, a potent and selective inhibitor of type I fatty acid synthase with central exposure.
Bioorganic & medicinal chemistry letters (2011-08-30)
Jörg T Kley, Jürgen Mack, Bradford Hamilton, Stefan Scheuerer, Norbert Redemann
ABSTRACT
Based on a high-throughput screen, cyclopentanecarboxanilides were identified as a new chemotype of non-covalent inhibitors of type I fatty acid synthase (FAS). Starting from initial hits we aimed at generating a tool compound suitable for the in vivo validation of FAS as a therapeutic target. Optimisation yielded BI 99179 which is characterised by high potency, remarkably high selectivity and significant exposure (both peripheral and central) upon oral administration in rats.
Sigma-Aldrich
BI 99179, ≥98% (HPLC)