Merck
  • SMaRT lncRNA controls translation of a G-quadruplex-containing mRNA antagonizing the DHX36 helicase.
SMaRT lncRNA controls translation of a G-quadruplex-containing mRNA antagonizing the DHX36 helicase.
EMBO reports (2020-04-28)
Julie Martone, Davide Mariani, Tiziana Santini, Adriano Setti, Sama Shamloo, Alessio Colantoni, Francesca Capparelli, Alessandro Paiardini, Dacia Dimartino, Mariangela Morlando, Irene Bozzoni
ABSTRACT
Guanine-quadruplexes (G4) included in RNA molecules exert several functions in controlling gene expression at post-transcriptional level; however, the molecular mechanisms of G4-mediated regulation are still poorly understood. Here, we describe a regulatory circuitry operating in the early phases of murine muscle differentiation in which a long non-coding RNA (SMaRT) base pairs with a G4-containing mRNA (Mlx-γ) and represses its translation by counteracting the activity of the DHX36 RNA helicase. The time-restricted, specific effect of lnc-SMaRT on the translation of Mlx-γ isoform modulates the general subcellular localization of total MLX proteins, impacting on their transcriptional output and promoting proper myogenesis and mature myotube formation. Therefore, the circuitry made of lnc-SMaRT, Mlx-γ, and DHX36 not only plays an important role in the control of myogenesis but also unravels a molecular mechanism where G4 structures and G4 unwinding activities are regulated in living cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human POMT1
Sigma-Aldrich
MISSION® esiRNA, targeting human DHX36
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution