Merck
  • Supplementation with a Carob (Ceratonia siliqua L.) Fruit Extract Attenuates the Cardiometabolic Alterations Associated with Metabolic Syndrome in Mice.
Supplementation with a Carob (Ceratonia siliqua L.) Fruit Extract Attenuates the Cardiometabolic Alterations Associated with Metabolic Syndrome in Mice.
Antioxidants (Basel, Switzerland) (2020-04-25)
María de la Fuente-Fernández, Daniel González-Hedström, Sara Amor, Antonio Tejera-Muñoz, Nuria Fernández, Luis Monge, Paula Almodóvar, Laura Andrés-Delgado, Luis Santamaría, Marin Prodanov, Antonio Manuel Inarejos-García, Angel Luis García-Villalón, Miriam Granado
ABSTRACT
The incidence of metabolic syndrome (MetS) is increasing worldwide which makes necessary the finding of new strategies to treat and/or prevent it. The aim of this study was to analyze the possible beneficial effects of a carob fruit extract (CSAT+®) on the cardiometabolic alterations associated with MetS in mice. 16-week-old C57BL/6J male mice were fed for 26 weeks either with a standard diet (chow) or with a diet rich in fats and sugars (HFHS), supplemented or not with 4.8% of CSAT+®. CSAT+® supplementation reduced blood glucose, Homeostatic Model Assessment of Insulin Resistance (HOMA-IR) and circulating levels of total cholesterol, low-density lipoprotein (LDL) cholesterol (LDL-c), insulin, and interleukin-6 (IL-6). In adipose tissue and skeletal muscle, CSAT+® prevented MetS-induced insulin resistance, reduced macrophage infiltration and the expression of pro-inflammatory markers, and up-regulated the mRNA levels of antioxidant markers. Supplementation with CSAT+® prevented MetS-induced hypertension and decreased the vascular response of aortic rings to angiotensin II (AngII). Moreover, treatment with CSAT+® attenuated endothelial dysfunction and increased vascular sensitivity to insulin. In the heart, CSAT+® supplementation reduced cardiomyocyte apoptosis and prevented ischemia-reperfusion-induced decrease in cardiac contractility. The beneficial effects at the cardiovascular level were associated with a lower expression of pro-inflammatory and pro-oxidant markers in aortic and cardiac tissues.
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, clone AW24 | 04-796, clone AW24, from rabbit