Merck
  • Engineered PLGA nano- and micro-carriers for pulmonary delivery: challenges and promises.
Engineered PLGA nano- and micro-carriers for pulmonary delivery: challenges and promises.
The Journal of pharmacy and pharmacology (2012-08-14)
Francesca Ungaro, Ivana d'Angelo, Agnese Miro, Maria I La Rotonda, Fabiana Quaglia
ABSTRACT
The aim of this review is to summarize the current state-of-the-art in poly(lactic-co-glycolic acid) (PLGA) carriers for inhalation. It presents the rational of use, the potential and the recent advances in developing PLGA microparticles and nanoparticles for pulmonary delivery. The most promising particle engineering strategies are discussed, highlighting the advantages along with the major challenges for researchers working in this field. Biodegradable polymer carriers, such as PLGA particles, may permit effective protection and long-term delivery of the inhaled drug and, when adequately engineered, its efficient transport to the target. The carrier can be designed for inhalation on the basis of several strategies through the adequate combination of available particle technologies and excipients. In so doing, the properties of PLGA particles can be finely tuned at micro-size and nano-size level to fulfill specific therapeutic needs. This means not only to realize optimal in vitro/in vivo lung deposition of the formulation, which is still crucial, but also to control the fate of the drug in the lung after particle landing. Although many challenges still exist, PLGA carriers may be highly beneficial and present a new scenario for patients suffering from chronic lung diseases and for pharmaceutical companies working to develop novel inhaled products.
Sigma-Aldrich
NanoFabTx PLGA-nano, for synthesis of 100 and 200 nm particles