Merck
  • Main Chain-Type Block Copolymers through Atom Transfer Radical Polymerization from Double-Decker-Shaped Polyhedral Oligomeric Silsesquioxane Hybrids.
Main Chain-Type Block Copolymers through Atom Transfer Radical Polymerization from Double-Decker-Shaped Polyhedral Oligomeric Silsesquioxane Hybrids.
Polymers (2020-02-23)
Wei-Cheng Chen, Yu-Hsuan Tsao, Chih-Feng Wang, Chih-Feng Huang, Lizong Dai, Tao Chen, Shiao-Wei Kuo
ABSTRACT
In this study, we synthesized two main chain-type block copolymers featuring hydrogen bond donor and acceptor segments through atom transfer radical polymerization (ATRP) using a bifunctionalized polyhedral oligomeric silsesquioxane (POSS) nanoparticle as the initiator. Hydrosilylation of vinylbenzyl chloride at the two corners of a double-decker silsesquioxane (DDSQ) provided the bifunctionalized benzyl chloride initiator VBC-DDSQ-VBC, which we applied as a platform to prepare a main chain-type polystyrene homopolymer (PS-DDSQ-PS), the diblock copolymer poly(styrene-b-4-vinylpyridine) (P4VP-b-PS-DDSQ-PS-b-P4VP), and the diblock copolymer poly(styrene-b-tert-butoxystyrene) (PtBuOS-b-PS-DDSQ-PS-b-PtBuOS) through sequential ATRP. Selective hydrolysis of the tert-butoxyl units of PtBuOS-b-PS-DDSQ-PS-b-PtBuOS yielded the strongly hydrogen bonding diblock copolymer poly (styrene-b-vinylphenol) (PVPh-b-PS-DDSQ-PS-b-PVPh). We used Fourier transfer infrared spectroscopy, nuclear magnetic resonance spectroscopy, size exclusion chromatography, differential scanning calorimetry, mass-analyzed laser desorption ionization mass spectrometry, and transmission electron microscopy to investigate the chemical structures, thermal behavior, and self-assembled nanostructures formed by these main chain-type block copolymers based on DDSQ.
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
4-tert-Butoxystyrene, 99%, contains 200 ppm 4-tert-butylcatechol as inhibitor