Merck
  • Effects of the anti-neoplastic agent ET-18-OCH3 and some analogs on the biophysical properties of model membranes.
Effects of the anti-neoplastic agent ET-18-OCH3 and some analogs on the biophysical properties of model membranes.
International journal of pharmaceutics (2006-04-21)
Alejandro Torrecillas, J Daniel Aroca-Aguilar, Francisco J Aranda, Consuelo Gajate, Faustino Mollinedo, Senena Corbalán-García, Ana de Godos, Juan C Gómez-Fernández
ABSTRACT
The effect of 1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine (ET-18-OCH(3), edelfosine), and six other analog asymmetric phosholipids on the physical properties of 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) model membranes was studied using differential scanning calorimetry (DSC), (31)P-nuclear magnetic resonance ((31)P NMR) and X-ray diffraction. DSC data revealed that, at concentrations of 40mol% and higher, a new type of mixtures with higher T(c) and narrower transitions appeared with all the asymmetric lipids studied. At very high concentrations of these lipids (50-80 mol%), destabilization was observed in the systems probably because of the formation of micelles or small vesicles. In all cases, the asymmetric lipids at concentrations of 40 mol% induced the formation of interdigitated structures in the lamellar gel phase, as deduced from X-ray diffraction. The asymmetric phospholipids were also added to 1,2-dielaidoyl-sn-glycero-3-phosphoethanolamine (DEPE) model membranes and DSC data revealed that the lipids primarily affected transition from the lamellar gel (L(beta)) to the lamellar liquid crystalline (L(alpha)) phase in two aspects: the transition temperature was reduced, and the transition itself became broader and smaller. The lamellar liquid crystalline (L(alpha)) to inverted hexagonal phase (H(II)) transition was also affected, as detected by DSC and (31)P NMR data. Increasing concentrations of the asymmetric lipids reduced the formation of inverted hexagonal phases, which were completely inhibited in the case of ET-18-OCH(3). Since these compounds have been shown to have important biological actions through the plasma membrane, these results may help to understand the mechanism of action of these compounds. In addition these asymmetric lipids were tested for their capacity to induce cell apoptosis, and only ET-18-OCH(3) was found to have a clear effect, thus suggesting that the apoptotic effect is not exerted through changes in the biophysical properties of model membranes.
Product Description
Avanti
16:0-02:0 PC, Avanti Research - A Croda Brand 880622C