Merck
  • Copolymerization of single-cell nucleic acids into balls of acrylamide gel.
Copolymerization of single-cell nucleic acids into balls of acrylamide gel.
Genome research (2019-11-16)
Siran Li, Jude Kendall, Sarah Park, Zihua Wang, Joan Alexander, Andrea Moffitt, Nissim Ranade, Cassidy Danyko, Bruno Gegenhuber, Stephan Fischer, Brian D Robinson, Herbert Lepor, Jessica Tollkuhn, Jesse Gillis, Eric Brouzes, Alex Krasnitz, Dan Levy, Michael Wigler
ABSTRACT
We show the use of 5'-Acrydite oligonucleotides to copolymerize single-cell DNA or RNA into balls of acrylamide gel (BAGs). Combining this step with split-and-pool techniques for creating barcodes yields a method with advantages in cost and scalability, depth of coverage, ease of operation, minimal cross-contamination, and efficient use of samples. We perform DNA copy number profiling on mixtures of cell lines, nuclei from frozen prostate tumors, and biopsy washes. As applied to RNA, the method has high capture efficiency of transcripts and sufficient consistency to clearly distinguish the expression patterns of cell lines and individual nuclei from neurons dissected from the mouse brain. By using varietal tags (UMIs) to achieve sequence error correction, we show extremely low levels of cross-contamination by tracking source-specific SNVs. The method is readily modifiable, and we will discuss its adaptability and diverse applications.
Sigma-Aldrich
1H,1H,2H,2H-Perfluoro-1-octanol, 97%
Sigma-Aldrich
Acrylamide/Bis-acrylamide, 40% solution, BioReagent, suitable for electrophoresis, 19:1
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei EZ Prep, sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
Fluorinert FC-40, immiscible fluorocarbon oil
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell