Merck
Function and regulation of cullin-RING ubiquitin ligases.
Nature reviews. Molecular cell biology (2005-02-03)
Matthew D Petroski, Raymond J Deshaies
ABSTRACT
Cullin-RING complexes comprise the largest known class of ubiquitin ligases. Owing to the great diversity of their substrate-receptor subunits, it is possible that there are hundreds of distinct cullin-RING ubiquitin ligases in eukaryotic cells, which establishes these enzymes as key mediators of post-translational protein regulation. In this review, we focus on the composition, regulation and function of cullin-RING ligases, and describe how these enzymes can be characterized by a set of general principles.