Merck
  • Heterogeneity of osteosarcoma cell lines led to variable responses in reprogramming.
Heterogeneity of osteosarcoma cell lines led to variable responses in reprogramming.
International journal of medical sciences (2014-08-30)
Pei Feng Choong, Hui Xin Teh, Hoon Koon Teoh, Han Kiat Ong, Kong Bung Choo, Shigeki Sugii, Soon Keng Cheong, Tunku Kamarul
ABSTRACT
Four osteosarcoma cell lines, Saos-2, MG-63, G-292 and U-2 OS, were reprogrammed to pluripotent state using Yamanaka factors retroviral transduction method. Embryonic stem cell (ESC)-like clusters started to appear between 15 to 20 days post transduction. Morphology of the colonies resembled that of ESC colonies with defined border and tightly-packed cells. The reprogrammed sarcomas expressed alkaline phosphatase and pluripotency markers, OCT4, SSEA4, TRA-1-60 and TRA-1-81, as in ESC up to Passage 15. All reprogrammed sarcomas could form embryoid body-like spheres when cultured in suspension in a low attachment dish for up to 10 days. Further testing on the directed differentiation capacity of the reprogrammed sarcomas showed all four reprogrammed sarcoma lines could differentiate into adipocytes while reprogrammed Saos-2-REP, MG-63-REP and G-292-REP could differentiate into osteocytes. Among the 4 osteosarcoma cell lines, U-2 OS reported the highest transduction efficiency but recorded the lowest reprogramming stability under long term culture. Thus, there may be intrinsic differences governing the variable responses of osteosarcoma cell lines towards reprogramming and long term culture effect of the reprogrammed cells. This is a first report to associate intrinsic factors in different osteosarcoma cell lines with variable reprogramming responses and effects on the reprogrammed cells after prolonged culture.
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, FITC conjugate, Chemicon®, from rabbit