Merck
  • Inhibition of smooth muscle contraction and ARF6 activity by the inhibitor for cytohesin GEFs, secinH3, in the human prostate.
Inhibition of smooth muscle contraction and ARF6 activity by the inhibitor for cytohesin GEFs, secinH3, in the human prostate.
American journal of physiology. Renal physiology (2017-09-01)
Annika Herlemann, Patrick Keller, Melanie Schott, Alexander Tamalunas, Anna Ciotkowska, Beata Rutz, Yiming Wang, Qingfeng Yu, Raphaela Waidelich, Frank Strittmatter, Christian G Stief, Christian Gratzke, Martin Hennenberg
ABSTRACT
Prostate smooth muscle contraction is critical for etiology and treatment of male lower urinary tract symptoms (LUTS) and is promoted by small monomeric GTPases (RhoA and Rac). GTPases may be activated by guanosine nucleotide exchange factors (GEFs). GEFs of the cytohesin family may indirectly activate Rac, or ADP ribosylation factor (ARF) GTPases directly. Here we investigated the expression of cytohesin family GEFs and effects of the cytohesin inhibitor Sec7 inhibitor H3 (secinH3) on smooth muscle contraction and GTPase activities in human prostate tissues. Of all four cytohesin isoforms, cytohesin-1 and -2 showed the highest expression in real-time PCR. Western blot and fluorescence staining suggested that cytohesin-2 may be the predominant isoform in prostate smooth muscle cells. Contractions induced by norepinephrine, the α1-adrenoceptor agonist phenylephrine, the thromboxane A2 analog U-46619 , and endothelin-1 and -3, as well as neurogenic contractions induced by electric field stimulation (EFS), were reduced by secinH3 (30 µM). Inhibition of EFS-induced contractions appeared to have efficacy similar to that of inhibition by the α1-adrenoceptor antagonist tamsulosin (300 nM). Combined application of secinH3 plus tamsulosin caused larger inhibition of EFS-induced contractions than tamsulosin alone. Pull-down assays demonstrated inhibition of the small monomeric GTPase ARF6 by secinH3, but no inhibition of RhoA or Rac1. In conclusion, we suggest that a cytohesin-ARF6 pathway takes part in smooth muscle contraction. This may open attractive new possibilities in medical treatment of male LUTS.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Rabbit Anti-Goat IgG Antibody, FITC conjugate, Chemicon®, from rabbit