Merck
  • The proteasome inhibitor bortezomib induces apoptosis in human retinoblastoma cell lines in vitro.
The proteasome inhibitor bortezomib induces apoptosis in human retinoblastoma cell lines in vitro.
Investigative ophthalmology & visual science (2007-09-28)
Vassiliki Poulaki, Constantine S Mitsiades, Vassiliki Kotoula, Joseph Negri, Douglas McMillin, Joan W Miller, Nicholas Mitsiades
ABSTRACT
To evaluate the potential of proteasome inhibitors, a novel class of antitumor agents, for the treatment of retinoblastoma. The proteasome inhibitor bortezomib (PS-341, Velcade; Millennium Pharmaceuticals, Cambridge, MA), approved by the US Food and Drug Administration for the treatment of multiple myeloma, is being studied for the treatment of several other malignancies. Among other effects, it inactivates the transcription factor nuclear factor-kappaB (NF-kappaB) by blocking the degradation of its inhibitor, IkappaB. NF-kappaB, which is constitutively active in human retinoblastoma cells and promotes their survival, represents a therapeutic target for patients with this malignancy. The authors evaluated the effect of bortezomib on the retinoblastoma cell lines Y79 and WERI-Rb1 in vitro using a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay, flow cytometry with propidium iodide, gene expression profiling, RT-PCR, and immunoblotting. Bortezomib induced caspase-dependent apoptosis in both retinoblastoma cell lines at clinically achievable concentrations. Bortezomib upregulated heat-shock proteins, other stress-response proteins, proapoptotic molecules, cell-cycle regulators, transcription factors, cytokines, and several proteasome subunits and solute carrier proteins, whereas it downregulated antiapoptotic and adhesion molecules. Bortezomib also induced cleavage of caspases, Bid and poly(ADP-ribose) polymerase (PARP), and sensitized retinoblastoma cells to doxorubicin. Bortezomib induces a stress response and triggers caspase-dependent apoptosis in human retinoblastoma cells at clinically achievable concentrations. This study provides insight into the molecular mechanism(s) of the antitumor activity of bortezomib and a basis for future preclinical studies leading to clinical trials of bortezomib, alone or in combination with conventional chemotherapy, to improve patient outcomes in retinoblastoma.
Sigma-Aldrich
Caspase Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.
Sigma-Aldrich
Anti-Bcl-X Antibody, clone 2H12, clone 2H12, Chemicon®, from mouse